Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium phosphate dibasic, anhydrous ≥99.0% ACS

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synonyms: Sodium phosphate dibasicdi-Sodium hydrogen orthophosphateSodium hydrogen phosphate

0404-12KG 0404-1KG 0404-2.5KG 0404-500G 0404-50KG
97061-588CS 775.79 USD InStock
97061-588 97061-584 97061-586 97061-590 97061-582
Sodium phosphate dibasic, anhydrous ≥99.0% ACS
di-Sodium hydrogen phosphate


Formula: Na₂HPO₄
MW: 141.96 g/mol
Melting Pt: 250 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003496
CAS Number: 7558-79-4
EINECS: 231-448-7
REACH: 01-2119489797-11
Merck Index: 12,08805
Order Now
Specification Test Results
Chloride 0.002%
Heavy Metals (as Pb) = 0.001%
Insolubles = 0.01%
Iron = 0.002%
Loss on Drying @105°C = 0.2%
pH (5%, Water) @25°C 8.7 - 9.3
Purity = 99%
Sulfate = 0.005%

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty