Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Urea, high purity

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synonyms: Carbonyl diamideCarbamideCarbonyldiamideDiaminomethanone

0568-2.5KG 0568-10KG 0568-5KG 0568-500G 0568-12KG 0568-1KG
97061-920CS 482.06 USD InStock
97061-920 97061-918 97061-926 97061-916 97061-922 97061-924
Urea, high purity
Urea
Commonly used to denature nucleic acids for electrophoresis and to study the secondary and tertiary structure of proteins. Effective for maintaining DNA in a highly denatured state during sequencing protocols, urea is also useful for accelerating the activity of Proteinase K digestions. Effectively denatures RNA. For use in nucleic electrophoresis applications.


Formula: NH₂CONH₂
MW: 60.06 g/mol
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 132.7 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008022
CAS Number: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 12,10005
Order Now
Specification Test Results
Abs.@280nm (5M, Water) 0.05
Biuret Test Pass
Chloride = 0.0005%
Cyanate = 0.002%
DNase none detected
Heavy Metals (as Pb) = 0.0005%
Insolubles = 0.005%
Iron = 0.0001%
Melting Point 132°C - 135°C
Protease none detected
Purity = 99.5%
RNase none detected
Solubility (5M, Water) Pass

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty