Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium dihydrogen phosphate, anhydrous ≥98.0%, Reagent Grade

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synonyms: Sodium dihydrogen phosphateSodium phosphate monobasicSodium dihydrogen orthophosphateMonobasic Sodium PhosphateSodium dihydrogenphosphate (granular)

0571-100KG 0571-1KG 0571-500G 0571-2.5KG
97061-940CS 570.02 USD InStock
97061-940 97061-942 97061-938 97061-934
Sodium dihydrogen phosphate, anhydrous ≥98.0%, Reagent Grade
Sodium dihydrogen phosphate


Formula: NaH₂PO₄
MW: 119.98 g/mol
Melting Pt: ∼650 °C
Density: 0.9996 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003527
CAS Number: 7558-80-7
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806
Order Now
Specification Test Results
Heavy Metals (as Pb) 0.001%
Insolubles = 0.03%
Moisture (KF) = 0.3%
pH (1%, Water) @25°C 4.4 - 4.7
Purity = 98%

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty