Synonyms:
Sodium 3α,12α-dihydroxy-5β-cholanoate
, Deoxycholic acid sodium salt
, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt
0613-50G
0613-500G
0613-100G
97062-024CS
2385.81
USD
InStock
97062-024
97062-028
97062-026
Sodium deoxycholate, high purity
Sodium deoxycholate
Anionic - negatively-charged (carboxylates, sulfates or sulfonates in alkali metal form). Utilized for applications ranging from solubilizing membrane proteins in non-denaturing conditions to creating denaturing conditions during gel electrophoresis. Used for complete disruption of membranes.