Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium deoxycholate, high purity

Supplier: VWR
Warning
Synonyms: Sodium 3α,12α-dihydroxy-5β-cholanoateDeoxycholic acid sodium salt3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt

0613-50G 0613-500G 0613-100G
97062-024CS 2385.81 USD InStock
97062-024 97062-028 97062-026
Sodium deoxycholate, high purity
Sodium deoxycholate
Anionic - negatively-charged (carboxylates, sulfates or sulfonates in alkali metal form). Utilized for applications ranging from solubilizing membrane proteins in non-denaturing conditions to creating denaturing conditions during gel electrophoresis. Used for complete disruption of membranes.


Formula: C₂₄H₃₉NaO₄
MW: 414.56 g/mol
Melting Pt: 357…365 °C
Storage Temperature: Ambient
MDL Number: MFCD00064139
CAS Number: 302-95-4
EINECS: 206-132-7
Order Now
Specification Test Results
Arsenic = 0.001 %
Heavy Metals = 0.005 %
Loss on Drying = 5.0 %
Purity (Dry Basis) 99.0%
Sodium Cholate = 2.0 %
Solubility (10%, Water) Pass

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty