Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium sulfite, anhydrous ACS

Supplier: VWR

0628-500G 0628-2.5KG 0628-1KG
97062-056CS 719.97 USD InStock
97062-056 97062-058 97062-060
Sodium sulfite, anhydrous ACS
Sodium sulphite


Formula: Na₂SO₃
MW: 126.04 g/mol
Melting Pt: > 500 °C
Density: 2.63 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
EINECS: 231-821-4
REACH: 01-2119537420-49
Merck Index: 12,08831
Order Now
Specification Test Results
Chloride 0.02%
Free Acid Test pass
Heavy Metals (as Pb) = 0.001%
Insolubles = 0.005%
Iron = 0.001%
Purity = 98%
Titrable Free Base = 0.03 meq/g

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty