Would you like to log in again?
Bovine Rnase A (from Pancreas)
Supplier: Avantor
0675-1G 0675-250MG 0675-500MG
97062-170EA 1195.7 USD InStock
97062-170 97062-174 97062-172
Bovine Rnase A (from Pancreas)
Enzymes
Ribonuclease A is an endoribonuclease that efficiently hydrolyzes RNA contaminants in DNA preparations from tissue or bacterial cell cultures. RNase A is used for a variety of molecular biology applications, most commonly for the hydrolysis of RNA that contaminates DNA preparations. This enzyme is used during plasmid purification without nicking or degrading plasmid.
  • Useful during DNA purification
  • High purity
  • Available in a ready-to-use solution
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty