Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Sorbitol ≥96%, high purity

Supplier: VWR
Synonyms: SorbitolD(-)-Glucitol(Sorbitol)(2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol

0691-5KG 0691-500G
97062-204CS 458.28 USD InStock
97062-204 97062-206
D-(-)-Sorbitol ≥96%, high purity
D(-)-Sorbitol


Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 98…100 °C
Density: 1.524 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004708
CAS Number: 50-70-4
EINECS: 200-061-5
Merck Index: 12,08873
https://us-legacy.vwr.com/stibo/web/std.lang.all/97/21/2299721.jpg
Order Now
Specification Test Results
Arsenic 0.0003 %
Chloride 0.005 %
Heavy Metals 0.001 %
Moisture (KF) = 1 %
Purity (Anhydrous) = 96 %
Residue after Ignition 0.1 %
Sulfate 0.01 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty