Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium dihydrogen phosphate, anhydrous ACS

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synonyms: MonopotassiumphosphatePotassium dihydrogen orthophosphatePotassium phosphate monobasicMonobasic Potassium PhosphatePotassium orthophosphate monobasicPotassium dihydrogenorthophosphate

0781-50KG 0781-2.5KG 0781-500G 0781-12KG 0781-1KG
97062-352EA 1572.69 USD InStock
97062-352 97062-346 97062-344 97062-348 97062-350
Potassium dihydrogen phosphate, anhydrous ACS
Potassium dihydrogen phosphate


Formula: KH₂PO₄
MW: 136.09 g/mol
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: 253 °C
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 12,07829
Order Now
Specification Test Results
Chloride 0.001%
Heavy Metals = 0.001%
Insolubles = 0.01%
Iron = 0.002%
Loss on Drying = 0.2%
Nitrogen Compounds = 0.001%
pH (5%, Water) @25°C 4.1 - 4.5
Purity = 99%
Sodium = 0.005%
Sulfate = 0.003%

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty