Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Agarose, II™ for biotechnology

Supplier: VWR

0815-100G 0815-25G 0815-250G
97062-398CS 12626.92 USD InStock
97062-398 97062-396 97062-400
Agarose, II™ for biotechnology
Agarose
A low-melting agarose that melts between 62 to 68 °C and remains liquid for several hours at 37 °C. Ideal for purification of macromolecules from agarose slices and for in-gel enzymatic manipulations.


Melting Pt: 60 - 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/43/75/23914375.jpg
Order Now
Specification Test Results
DNase NONE
EEO (-Mr) ≤0.12
Gel Strength (1.0%) ≥250 g/cm²
Gelling Range (1.0%) 24 - 29 °C
Melting Range 62 - 68 °C
Moisture < 10 %
Protease NONE
RNase NONE
Sulfate ≤0.1 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty