Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium chloride, anhydrous ≥96% ACS

Supplier: VWR
Warning
Synonyms: Calcium dichlorideChloro calciumCaCl2

1B1110-500G 1B1110-1KG 1B1110-2.5KG
97062-586CS 562.32 USD InStock
97062-586 97062-588 97062-590
Calcium chloride, anhydrous ≥96% ACS
Calcium chloride


Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 772 °C
Density: 2.174 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 13,01660
Order Now
Specification Test Results
Purity 96%
Titrable Free Base = 0.006 meq/g

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty