Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-(N-Morpholino)ethanesufonic acid (MES) monohydrate free acid, Ultrapure

Supplier: VWR
Warning
Synonyms: 2-Morpholinoethanesulphonic acid hydrate2-(N-Morpholino)ethanesulfonic acid monohydrate2-(N-Morpholino)ethanesulphonic acid monohydrate2-Morpholinoethanesulfonic acid hydrate

E169-100G E169-500G E169-250G
97062-640CS 7191.74 USD InStock
97062-640 97062-638 97062-636
β-(N-Morpholino)ethanesufonic acid (MES) monohydrate free acid, Ultrapure
β-(N-Morpholino)ethanesulphonic acid (MES) monohydrate
Biological buffers are useful for cell culture in vitro, enzyme assays, and some electrophoretic applications at physiological pH.


Formula: C₆H₁₃NO₄S·1H₂O
MW: 213.25 g/mol
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149409
CAS Number: 145224-94-8
EINECS: 224-632-3
Merck Index: 13,05929
Order Now
Specification Test Results
Abs.@260nm (1M, Water) 0.02
Abs.@280nm (1M, Water) 0.02
DNase none detected
Appearance White crystalline powder
Moisture (KF) 10 %
pKa @25°C 5.9 - 6.3
Protease none detected
Purity (Dry Basis,Titration) 99 %
RNase none detected
Solubility (1M, Water) Pass

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty