Would you like to log in again?
Proteinase K
Supplier: Avantor
E195-5ML E195-25ML
97062-670EA 265.14 USD InStock
97062-670 97062-668
Proteinase K
Enzymes
Proteinase K is a non-specific serine protease with a molecular weight of approximately 18 kDa.
This enzyme exhibits high stability and activity in the presence of SDS, EDTA, and urea, as well as over a wide pH range. Proteinase K is useful for the inactivation of nucleases during the isolation of DNA and RNA. Active over a broad pH range.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty