Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water for biotechnology nuclease-free, 0.2 µm filtered

Supplier: VWR
Synonyms: Dihydrogen oxide

Filtered through a 0.2 µm filter.

E476-100ML E476-500ML E476-1L
97062-792EA 76.52 USD InStock
97062-792 97062-794 97062-790
Water for biotechnology nuclease-free, 0.2 µm filtered
Water


Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 12,10175
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty