Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Biotechnology Grade Phosphate Buffered Saline

Supplier: Avantor

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
E504-100ML E504-500ML
97062-818EA 63.49 USD InStock
97062-818 97062-816
VWR® Biotechnology Grade Phosphate Buffered Saline
Buffers Electrophoresis Buffers Washing Buffers
1X Solution Composition:137 mM NaCl, 2.7 mM KCl, 12 mM Phosphate buffer

  • Does not contain magnesium or calcium
  • Solution is 0.2 µm filtered into bottles and autoclaved
  • Conductivity at 25°C (1X PBS): Lot Dependent
  • pH at 25°C (1X PBS): 7.3 – 7.4
  • Sterility(Milliflex):Pass.
  • DNase, none detected

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty