Would you like to log in again?
VWR® Stripping Buffers
Supplier: Avantor
VWR®
N552-500ML N552-1L N592-500ML N552-15ML
97064-220EA 140.09 USD InStock
97064-220 97064-242 97063-212 97064-218
VWR® Stripping Buffers
Buffers Electrophoresis Buffers Stripping Buffers
VWR® offers three stripping buffers to accommodate the wide range of conditions that may be needed to remove different antibodies from blots.
  • ReView™ Buffer Solution
  • Save money and time
  • Compare detection of different antibodies on a single blot
  • Gentle ReView™ Buffer
  • Odor-free, room-temperature procedure—No hood needed
  • Minimize loss of valuable antigen
  • Vantage ReView™ Stripping B
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty