Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Octyl-β-D-thioglucopyranoside

Supplier: VWR
Synonyms: (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(octylthio)tetrahydro-2H-pyran-3,4,5-triolN-Octyl-1-thio-Beta-D-glucopyranosideOTG1-S-Octyl-β-D-thioglucopyranosideOctyl-thioglucoside

J575-1G
97063-228EA 113.07 USD InStock
97063-228
Octyl-β-D-thioglucopyranoside
Octyl β-D-thioglucopyranoside
Non-ionic - containing no charge (polyoxyethylenes or alkyl glucosides). Non-ionic detergents are suited for solubilizing membrane proteins without altering biological activity. These non-denaturing, gentler detergents are ideal for studying the conformat


Formula: C₁₄H₂₈O₅S
MW: 308.44 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00012189
CAS Number: 85618-21-9
Order Now
Specification Test Results
Purity = 98%
Solubility (5%, Water) Pass
Specific Rotation (1%, MeOH) (- 55°) - (-49°)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty