Would you like to log in again?
Leupeptin
Supplier: Avantor
J580-5MG J580-25MG
97063-234EA 93.42 USD InStock
97063-234 97064-236
Leupeptin
Proteins and Peptides
A reversible inhibitor of cysteine proteases and serine proteases resembling trypsin
  • Reversible inhibitor
  • Ultra pure grade
Inhibits trypsin, plasmin, papain, kallikrein, thrombin and cathepsin A and B.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty