Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

IPTG (isopropyl-β-D-thiogalactopyranoside), Ultrapure

Supplier: VWR
Warning
Synonyms: (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol

N679-10G
97063-282EA 683.39 USD InStock
97063-282
IPTG (isopropyl-β-D-thiogalactopyranoside), Ultrapure
IPTG (Isopropyl-β-D-thiogalactopyranoside)
Non-animal origin (NAO) IPTG is recommended lacZ selection applications where animal derived products are prohibited. NAO IPTG is free of animal products and dioxane free. Purity is greater than 98%.


Formula: C₉H₁₈O₅S
MW: 238.31 g/mol
MDL Number: MFCD00063273
CAS Number: 367-93-1
Order Now
Specification Test Results
Abs.@300nm (5%, Water) 0.15
Abs.@400nm (5%, Water) 0.06
Dioxane Test Pass
Appearance White crystalline powder
Melting Point 110°C - 114°C
Moisture (KF) 1 %
pH (5%, Water)@25°C 5 - 7
Purity 98 %
Specific Rotation -34.5° - -28.5°

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty