Would you like to log in again?
VWR® TRIS-Borate-EDTA Buffer, 5X Concentrate, Ultra Pure Grade
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
J885-4L J885-1L J885-5L
97063-432EA 49.23 USD InStock
97063-432 97063-438 97063-436
VWR® TRIS-Borate-EDTA Buffer, 5X Concentrate, Ultra Pure Grade
Buffers Electrophoresis Buffers Running Buffers
TBE is optimized for DNA electrophoresis techniques using both acrylamide and agarose
  • High Buffering Capacity
  • High Ionic Strength
  • Permits Extended Electrophoresis Runs with no Buffer Recirculation
  • Convenient Packaging Options Minimize Weighing and Mixing
After dilution the composition of (1X) Solution contains 0.089 M Tris Base, 0.089 M Borate and 0.002 M EDTA, Disodium Salt, Dihydrate; final pH 8.3.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty