Would you like to log in again?
VWR® 2XYT Medium Broth
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
J902-500G J902-100G
97063-440CS 542.31 USD InStock
97063-440 97063-442
VWR® 2XYT Medium Broth
Media for Microbiology Dehydrated Media for Microbiology
2XYT Medium Broth powder is used to prepare rich broth for the propagation of M13 bacteriophage.
  • Save time using premixed and ready-to-use media
  • Homogenous and high quality powders
  • Light beige, free flowing powder
31g of powder when dissolved per liter of deionized water will yield formulation- 16 grams/L of cesein peptone, 10 grams/L of casamino acids and 5 grams/L of sodium chloride.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty