Would you like to log in again?
VWR® DNA Molecular Weight Markers, 100 bp Ladder
Supplier: Avantor
VWR®
N550-300UL K180-250UL N856-300UL
97063-350EA 161.66 USD InStock
97063-350 97063-488 97063-208
VWR® DNA Molecular Weight Markers, 100 bp Ladder
Electrophoresis Reagents Molecular Weight Markers
13 fragments ranging from 100 to 3000 bp for easy band identification.
  • Easy band identification
  • Available with loading dye
Also available pre-mixed with loading dye or with EZ-Vision® One DNA dye.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty