Would you like to log in again?
VWR® 50X TRIS-Acetate-EDTA Buffer, Ultra Pure Grade
Supplier: Avantor
VWR®
K915-4L K915-1.6L K915-5L
97063-692EA 234.24 USD InStock
97063-692 97063-698 97063-696
VWR® 50X TRIS-Acetate-EDTA Buffer, Ultra Pure Grade
Buffers Electrophoresis Buffers Running Buffers
TAE is an extensively used buffer for agarose gel electrophoresis applications requiring high resolution and separation of high molecular weight, double-stranded DNA
TAE is more compatible with in-gel manipulations and band recovery procedures than a TBE buffer. A single strength (1X) solution contains 0.04M Tris-Acetate and 0.001M EDTA with a final pH of 8.0. Low ionic strength. Better resolution of large DNA fragments (>12kbDNA).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty