Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Guanidinium hydrochloride 8 M in aqueous solution, Biotechnology Grade

Supplier: VWR
Synonyms: Guanidinium chlorideGuanidine hydrochlorideGuanidine chlorideGuanadine hydrochloride

M113-4L
97063-774EA 1739.69 USD InStock
97063-774
Guanidinium hydrochloride 8 M in aqueous solution, Biotechnology Grade
Guanidinium hydrochloride
A concentrated solution prepared with Biotechnology Grade powder for convenient use in nucleic acid and protein purification procedures.


Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
MDL Number: MFCD00013026
CAS Number: 50-01-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/24/2297424.jpg
Order Now
Specification Test Results
Protease none detected

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty