Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Coomassie brilliant blue G-250, Proteomics Grade

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Warning
Synonyms: Xylene Brilliant Cyanine GC.I. 42655Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulphonatobenzyl)amino]-2'-methylbenzhydrylene]-3-methylcyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulphonatobenzyl)ammonium monosodium saltSodium 3-(((4-((4-((4-ethoxyphenyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)-2-methylcyclohexa-2,5-dien-1-ylidene)methyl)-3-methylphenyl)(ethyl)amino)methyl)benzenesulphonateServa Blue GHydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]-2'-methylbenzhydrylene]-3-methylcyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulfonatobenzyl)ammonium monosodium saltSodium 3-(((4-((4-((4-ethoxyphenyl)amino)phenyl)(4-(ethyl(3-sulfonatobenzyl)iminio)-2-methylcyclohexa-2,5-dien-1-ylidene)methyl)-3-methylphenyl)(ethyl)amino)methyl)benzenesulfonateAcid Blue 90Brilliant indocyanine GCoomassie Blue G 250Brilliant Blue GBrillant blue G

M140-50G M140-10G M140-25G
97063-852EA 75.63 USD InStock
97063-852 97063-850 97063-854
Coomassie brilliant blue G-250, Proteomics Grade
Coomassie brilliant blue G-250


Formula: C₄₇H₄₈N₃NaO₇S₂
MW: 854.04 g/mol
Density: >1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078482
CAS Number: 6104-58-1
EINECS: 228-058-4
Order Now
Specification Test Results
Em (595nm, Water) 36300
Protease none detected
Protein Staining Pass
Appearance Blue to red crystalline powder

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty