Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzamidine hydrochloride hydrate ≥98.0% (by anhydrous basis), Proteomics Grade

Supplier: VWR
Warning
Synonyms: Phenylamidine hydrochloride hydrateBenzenecarboximidamide hydrochloride hydrateBenzamidinium chloride hydrateBenzene-1-carboximidamide hydrochloride hydrate

M141-100G
97063-856CS 3890.76 USD InStock
97063-856
Benzamidine hydrochloride hydrate ≥98.0% (by anhydrous basis), Proteomics Grade
Benzamidine hydrochloride hydrate
A protease inhibitor of arginine-selective enzymes like trypsin, kallikrein and thrombin.

  • Product is animal-free

Extracted proteins are exposed to the threat of degradation by proteases from the moment cells or tissues are lysed. Proteases present in the living cell are released from their regulated environment into the extract where they may freely degrade their targets, resulting in drastically reduced protein yield and sample
quality. Complex protein solutions are typically treated with a mixture of several protease inhibitors in order to preserve the native state of the proteins.


Formula: C₆H₅C(=NH)NH₂·HCl·H₂O
MW: 174.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066285
CAS Number: 206752-36-5
EINECS: 216-795-4
Order Now
Specification Test Results
Em (Lambda Max, MEOH, anhy) 9500
Heavy Metals (as Pb) 0.005 %
Identification (IR) PASS
Melting Point 77 - 87 °C
Purity (anhydrous) 98.0 %
Reassay Date REPORT
Solubility (1%, Water) NONE

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty