Would you like to log in again?
VWR® Pepstatin
Supplier: Avantor
M183-5MG
97063-936EA 85.3 USD InStock
97063-936
VWR® Pepstatin
Proteins and Peptides
Product is animal-free
  • Activity: Working range 0.7 µg/ml (1 µM)
Extracted proteins are exposed to the threat of degradation by proteases from the moment cells or tissues are lysed. Proteases present in the living cell are released from their regulated environment into the extract where they may freely degrade their targets, resulting in drastically reduced protein yield and sample
quality. Complex protein solutions are typically treated with a mixture of several protease inhibitors in order to preserve the native state of the proteins.
An aspartic acid protease inhibitor of pepsin, renin and cathepsin D. Also an inhibitor of the HIV protease. Not water soluble.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty