Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Neutral red, Certifiable Grade

Supplier: VWR
Synonyms: N2,N2,7-Trimethylphenazine-2,8-diamine hydrochlorideToluylene redCI 500403-Amino-7-dimethylamino-2-methylphenazineN8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochlorideBasic Red 5

E895-100G
97064-080EA 313.25 USD InStock
97064-080
Neutral red, Certifiable Grade
Neutral red
A cationic azine dye slightly soluble in water and ethanol. Used as a weak nuclear stain and as a supravital stain of fresh peripheal blood.


Formula: C₁₅H₁₇ClN₄
MW: 288.78 g/mol
Melting Pt: 290 °C
Storage Temperature: Ambient
MDL Number: MFCD00012651
CAS Number: 553-24-2
EINECS: 209-035-8
Merck Index: 12,06571
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty