Would you like to log in again?
VWR® Agarose SFR™ Agarose SFR
Supplier: Avantor
Agarose SFR™
J234-100G J234-250G
97064-134CS 5364.09 USD InStock
97064-134 97064-136
VWR® Agarose SFR™ Agarose SFR
Electrophoresis Gels Agarose and Acrylamide
Agarose SFR™ (Super Fine Resolution) is a high resolution sieving agarose with excellent clarity. DNA bands differing in size by 2% can be resolved in the range of 200 bp to 1000 bp.
  • Super fine resolution - resolve 238 bp and 242 bp bands
  • Low melting point
  • Nuclease and protease-free
This agarose is suitable for the analysis of AFLPs (Amplified Fragment Length Polymorphisms), STRs (Short Tandem Repeats) and tetranucleotide repeats. The low melting temperature of Agarose SFR™ makes it an excellent medium for analytical and preparative electrophoresis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty