Would you like to log in again?
VWR® Pepstatin
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
J583-5MG J583-25MG
97063-246EA 70.41 USD InStock
97063-246 97064-248
VWR® Pepstatin
Proteins and Peptides
An aspartic acid protease inhibitor of pepsin, renin and cathepsin D
  • Aspartic acid
  • Not water soluble
  • Ultra pure grade
Also an inhibitor of the HIV protease. Not water soluble.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty