Would you like to log in again?
VWR Life Science Phenol-Free Total RNA Purification Kit
Supplier: Avantor
VWR®
N788-KIT
97064-308EA 617.3 USD InStock
97064-308
VWR Life Science Phenol-Free Total RNA Purification Kit
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
  • Save time
  • Reduce hazardous chemical use
  • Enhance recovery of total RNA including microRNA species
VWR’s Phenol-FreeTotal RNA Purification Kit provides a rapid method for the isolation and purification of total RNA from cultured animal cells, tissue samples, blood, bacteria, yeast, fungi, plants and viruses. The kit purifies all sizes of RNA, from large mRNA and ribosomal RNA down to microRNA (miRNA) and small interfering RNA (siRNA). The RNA is preferentially purified from other cellular components, such as proteins, without the use of phenol or chloroform. The purified RNA is of the highest integrity, and can be used in a number of downstream applications including real time PCR, reverse transcription PCR, Northern blotting, RNase protection and primer extension, and expression array assays.
Ordering information: Lysis Solution, 40 ml
Wash Solution, 22ml
Elution Solution, 6 ml
Mini Spin Columns, 50
Collection Tubes, 50
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty