Would you like to log in again?
VWR® Rapid Transfer Buffer, 10X
Supplier: Avantor
VWR®
N789-4L N789-1L
97064-312EA 161.48 USD InStock
97064-312 97064-314
VWR® Rapid Transfer Buffer, 10X
Buffers Electrophoresis Buffers Transfer Buffers
Rapid Transfer Buffer is a simple, single component system for quick and efficient transfer of proteins from SDS-PAGE gels to membranes for Western blotting applications
  • Fast, efficient transfer in 10–20 minutes
  • Utilizes standard transfer equipment
  • Non-hazardous formulation
  • Transfers to PVDF and nitrocellulose from multiple types of gels
Transfer is completed in 10 or 20 minutes using a standard semi-dry or wet transfer apparatus, respectively. Rapid Transfer Buffer is a methanol-free, non-hazardous formulation that is compatible with PVDF and nitrocellulose membranes. It is used for transfer of proteins from standard Laemmli gels, commercial pre-cast gels, and VWR’s NEXT GEL™. The transfer efficiency is equivalent to that observed when using a Tris-Glycine-Methanol transfer buffer.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty