Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) sulfate pentahydrate ACS

Supplier: VWR
Warning
Synonyms: Cupric sulfate pentahydrateCopper sulfate pentahydrateCopper sulphate pentahydrateCupric sulphate pentahydrate

0330-500G 0330-1KG 0330-2.5KG
97064-980CS 992.86 USD InStock
97064-980 97064-564 97065-032
Copper(II) sulfate pentahydrate ACS
Copper (II) sulphate pentahydrate


Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Density: 2.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3077
ADR: 9,III
REACH: 01-2119520566-40
Merck Index: 13,02682
Order Now
Specification Test Results
Calcium 0.005%
Chloride = 0.001%
Insolubles = 0.005%
Iron = 0.003%
Nickel = 0.005%
Nitrogen Compounds (as N) = 0.002%
Potassium = 0.01%
Purity 98% - 102%
Sodium = 0.02%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty