Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver nitrate ACS

Supplier: VWR
Danger
Synonyms: Silver mononitrateNitric acid silver saltLunar caustic

0377-25G 0377-100G
97064-580CS 6195.29 USD InStock
97064-580 97064-582
Silver nitrate ACS
Silver nitrate
Staining protein after electrophoretic separation. Also used to determine chloride ions in solution.


Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444 °C (1013 hPa)
Melting Pt: 210 °C
Density: 4.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1,II
REACH: 01-2119513705-43
Merck Index: 12,08661
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/45/16640645.jpg
Order Now
Specification Test Results
Appearance (ACS) Colorless or white solid.
Chloride = 0.0005%
Clarity of Solution Pass
Copper = 0.0002%
Free Acid Test Pass
Iron = 0.0002%
Lead = 0.001%
Purity = 99%
Substances not Precipitated by HCl = 0.01%
Sulfate = 0.002%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty