Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenol ≥99% ACS

Supplier: VWR
Danger
Synonyms: Carbolic acidHydroxybenzenePhenylalcohol

0735-250G 0735-500G
97064-666CS 686.93 USD InStock
97064-666 97064-668
Phenol ≥99% ACS
Phenol


Formula: C₆H₅OH
MW: 94.11 g/mol
Boiling Pt: 181.8 °C (1013 hPa)
Melting Pt: 40.8 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 81 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS Number: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1,II
Order Now
Specification Test Results
Clarity of Solution Pass
Freezing Range =40.5°C
Moisture (KF) = 0.5%
Purity = 99%
Residue After Evaporation = 0.05%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty