Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenol liquified ≥89.0%, Bioreagent

Supplier: VWR
Danger

E357-500ML
97064-752CS 583.2 USD InStock
97064-752
Phenol liquified ≥89.0%, Bioreagent
Phenol liquified
VWR's phenol products are manufactured from the purest crystalline phenol to ensure optimal quality.


Formula: C₆H₅OH
MW: 94.11 g/mol
Melting Pt: 40 °C…42 °C
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS Number: 108-95-2
EINECS: 203-632-7
UN: 2821
ADR: 6.1,II
Order Now
Specification Test Results
Distilling Range = 182.5° C
Nonvolatile Residue = 0.05 %
Purity (%) 89.0

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty