Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide, pellets, Proteomics Grade

Supplier: VWR
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

M137-1KG M137-500G
97065-008CS 370.1 USD InStock
97065-008 97064-896
Sodium hydroxide, pellets, Proteomics Grade
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C
Melting Pt: 318 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627
Order Now
Specification Test Results
Chloride 0.005%
DNase none detected
Heavy Metals 0.002 %
Iron = 0.001 %
Mercury = 0.1 ppm
Nickel = 0.001 %
Nitrogen Compounds = 0.001 %
Phosphate = 0.001 %
Potassium = 0.02 %
Protease none
Purity = 97 %
RNase none detected
Sodium Carbonate = 1 %
Sulfate = 0.003%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty