Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oxalic acid dihydrate, high purity

Supplier: VWR
Warning
Synonyms: Ethanedioic acid dihydrate

0326-1KG 0326-500G
97064-562CS 782.16 USD InStock
97064-562 97064-978
Oxalic acid dihydrate, high purity
Oxalic acid dihydrate


Formula: HO₂CCO₂H·2H₂O
MW: 126.07 g/mol
Boiling Pt: 149…160 °C (1013 hPa)
Melting Pt: 101 °C
Density: 1.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149102
CAS Number: 6153-56-6
EINECS: 205-634-3
UN: 3261
ADR: 8,III
REACH: 01-2119534576-33
Merck Index: 12,07043
https://us-legacy.vwr.com/stibo/web/std.lang.all/72/16/2297216.jpg
Order Now
Specification Test Results
Ash 0.2 %
Chloride 0.005 %
Insolubles 0.05 %
Iron 0.005 %
Purity 99 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty