Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium nitrate ACS

Supplier: VWR
Warning

0700-1KG
97065-038CS 414.33 USD InStock
97065-038
Potassium nitrate ACS
Potassium nitrate


Formula: KNO₃
MW: 101.1 g/mol
Melting Pt: 334 °C
Density: 2.109 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011409
CAS Number: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1,III
REACH: 01-2119488224-35
Merck Index: 12,07815
Order Now
Specification Test Results
Calcium 0.005%
Chloride = 0.002%
Heavy Metals (as Pb) = 0.0005%
Insolubles = 0.005%
Iodate = 0.0005%
Iron = 0.0003%
Magnesium = 0.002%
Nitrite = 0.001%
pH (5%, Water) @25°C 4.5 - 8.5
Phosphate = 0.0005%
Purity = 99%
Sodium = 0.005%
Sulfate = 0.003%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty