Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol, anhydrous ≥99.8%, Ultrapure

Supplier: VWR
Danger
Synonyms: Hydroxy methaneMethyl hydroxideMethyl alcoholMethylolCarbinolWood alcohol

0323-4L
97065-052CS 600.93 USD InStock
97065-052
Methanol, anhydrous ≥99.8%, Ultrapure
Methanol


Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6 °C
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 Subrisk 6.1,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg
Order Now
Specification Test Results
Moisture (KF) = 0.1 %
Purity = 99.8 %
Specific Gravity 0.783 - 0.791

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty