Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Lactic acid 85-90% ACS

Supplier: VWR
Danger
Synonyms: (S)-(+)-2-Hydroxypropionic acid(S)-2-Hydroxypropanoic acid(S)-(+)-Lactic acid(S)-(+)-2-Hydroxypropanoic acid

0349-4L 0349-1L
97064-984EA 43.4 USD InStock
97064-984 97065-054
L(+)-Lactic acid 85-90% ACS
L(+)-Lactic acid


Formula: C₃H₆O₃
MW: 90.08 g/mol
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: ∼53 °C
Density: ∼1.385 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00064266
CAS Number: 79-33-4
EINECS: 201-196-2
Merck Index: 13,05352
Order Now
Specification Test Results
Chloride 0.001%
Heavy Metals (as Pb) = 0.0005%
Iron = 0.0005%
Purity 85% -90%
Residue on Ignition = 0.02%
Substances Darkened by H2SO4 Pass
Sulfate = 0.002%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty