Would you like to log in again?
Corning® Dialyzed FBS, Corning
Supplier: Corning
Corning®
35-071-CV 35-071-CV
97065-302EA 1673.23 USD InStock
CA97065-302 97065-302
Corning® Dialyzed FBS, Corning
Sera (Serum)
As a natural product, fetal bovine serum contains a wide variety of biological molecules and compounds which can influence cell development
  • Sera is aseptically collected and processed in a cGMP facility
  • Each batch of raw serum is screened for endotoxin, residual hemoglobin concentration, and microbiological quality before being used in production
  • Batches of raw serum are pooled and continuously mixed prior to filtration to ensure consistent quality throughout the entire lot
  • Final lots are tested for endotoxin, hemoglobin, pH, mycoplasma, and growth promotion. Biochemical assays and virus testing are also available upon request
  • Premium FBS is sourced from U.S. herds
  • Dialyzed via a 10,000MW cutoff filter and dialyzed against normal saline
This serum is dialyzed against physiological saline to remove small (less than 10,000 MW) potential contaminants while preserving the biological integrity of the final product. An extra filtration step of 0.1 micron further ensures purity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty