Would you like to log in again?
Paq5000 DNA Polymerase, Agilent Technologies
Supplier: Agilent Technologies
Paq5000
600860 600680 600684 600864 600682 600682 600680 600872 600864 600684 600862 600860 600870 600872 600870
97066-514EA 181.9 USD InStock
CA97066-522 97066-514 CA97066-514 97066-520 97066-522 CA97066-520 99902-838 CA99902-838 CAST600680 ST600680 CA99904-872 97066-518 CA97066-516 99904-872 CA97066-518
Paq5000 DNA Polymerase, Agilent Technologies
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Paq5000 is capable of quickly and robustly amplifying up to 6 kB genomic DNA targets.
  • Equal to better yield with fast cycling conditions
  • Robust, cost-effective
  • Pre-optimized reaction buffers included
  • Low cost hot start formulation
With a shortened extension rate of 30 sec/kB and robust yields, Paq5000 is a Taq alternative that can be substituted easily into many Taq-based procedures.
Paq5000 includes the ArchaeMaxx factor, improving the yield of products by overcoming dUTP poisoning, which is caused by dUTP accumulation during PCR through dCTP deamination. Once incorporated, dU-containing DNA inhibits the enzyme and the ArchaeMaxx factor functions as a dUTPase, converting poisonous dUTP to harmless dUMP and inorganic pyrophosphate.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty