Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
GENbag Atmosphere Generators, bioMérieux
Supplier: bioMérieux
45532 45534
97067-038PK 114.53 USD InStock
97067-038 97067-042
GENbag Atmosphere Generators, bioMérieux
Incubators Standard Incubators
GENbag enables the incubation of up to 5 plates
Bacterial growth can be observed without opening, through the transparent bag
Ordering information: Kit Includes 20 airlight bags, 20 generators, and 2 dip seals.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty