Would you like to log in again?
Axygen® AxyPrep™ FragmentSelect-I Kit, Corning
Supplier: Corning
Axygen® AxyPrep™
MAG-FRAG-I-1 MAG-FRAG-I-250 MAG-FRAG-I-5 MAG-FRAG-I-50
10159-708CS 120.38 USD InStock
10159-708 97067-607 97067-615 97067-609
Axygen® AxyPrep™ FragmentSelect-I Kit, Corning
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The AxyPrep™ FragmentSelect-I Kit is optimized for Illumina next generation sequencing platforms and Life Technologies (SOLiD) fragment size selection needs to simplify the library construction workflow. The kit is designed for fragment size selection needs in the Roche/454’s DNA Genome sequencer library construction workflow.
  • Optimized fragment selection chemistries for Illumina, SOLiD, and Roche/454
  • Versatile and flexible sequential fragment size selection; no gels needed
  • Efficient removal of primer-dimers; no need for further clean-up steps
  • Manual and automation friendly
  • Scalable: Tube, 96-, and 384-well plate formats
  • No centrifugation or filtration required
Post DNA shearing, the library construction process for next generation sequencing mostly requires fragment selection regardless of the platform
The AxyPrep™ FragmentSelect-I Kits utilize a unique paramagnetic bead technology for quick high-throughput optimized DNA size selection suitable for various next generation sequencing platforms.
Obtaining high recovery post fragment selection is becoming an important contributor for the reduction of sequencing bias.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty