Would you like to log in again?
Axygen® Agarose LM, Molecular Biology Grade, Corning
Supplier: Corning
Axygen®
AGR-LM-1001 AGR-LM-501
97067-662CS 2464.77 USD InStock
97067-662 97067-666
Axygen® Agarose LM, Molecular Biology Grade, Corning
Electrophoresis Gels Agarose and Acrylamide
Axygen® Agarose Low Melting (LM) is a low melting agarose that allows for the recovery of undamaged nucleic acids below their denaturation temperature.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty