Would you like to log in again?
Axygen® Agarose MS, PCR Screening, Molecular Biology Grade, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Axygen®
AGR-MS-1001
97067-668CS 4205.38 USD InStock
97067-668
Axygen® Agarose MS, PCR Screening, Molecular Biology Grade, Corning
Electrophoresis Gels Agarose and Acrylamide
Axygen® Agarose Molecular Screening (MS) is a molecular screening agarose for improved resolution of DNA fragments with 500bp or less, especially primer-sized fragments.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty