Would you like to log in again?
HiPrep Sephacryl S-500 HR Columns
Supplier: Cytiva
HiPrep™
28935606 28935607 28-9356-07 28-9356-06
97067-774EA 1295.95 USD InStock
97067-774 CA97067-772 97067-772 CA97067-774
HiPrep Sephacryl S-500 HR Columns
Chromatography Columns
HiPrep 26/60 Sephacryl S-500 HR are prepacked gel filtration column for preparative separation of biomolecules from up to 13 ml sample volume.
  • Gel filtration columns for low to medium pressure ÄKTA systems
  • Well-established in industrial production
  • The HiPrep Sephacryl series offers reproducible separations over a wide molecular weight range
  • Convenient, prepacked HiPrep Sephacryl columns speed up protein purification
Sephacryl S-500 is used for separating polysaccarides, macromolecules with extended structures, and even small plasmids. Maximum recovery and minimum nonspecific adsorption with long-term chemical and physical stability resulting from the hydrophilic, rigid allyl dextran/bisacrylamide matrix.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty