Would you like to log in again?
HiScreen Capto Q ImpRes Columns, Cytiva
Supplier: Cytiva
HiScreen™
17-5470-15 17547015
97067-786EA 190.25 USD InStock
97067-786 CA97067-786
HiScreen Capto Q ImpRes Columns, Cytiva
Chromatography Columns
HiScreen Capto Q ImpRes columns are prepacked with Capto Q ImpRes ion exchange chromatography medium, and are part of the process development platform available from Cytiva.
  • Capto Q ImpRes is a strong anion exchange BioProcess medium, designed to meet the demands of modern large-scale manufacturers for fast, efficient, and cost-effective intermediate and polishing protein purification
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
The columns are an excellent choice for method optimization and parameter screening.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty