Would you like to log in again?
HiScreen MabSelect SuRe LX Columns, Cytiva
Supplier: Cytiva
HiScreen™ MabSelect SuRe™
17-5474-15 17547415
97067-788EA 1927.72 USD InStock
CA97067-788 97067-788
HiScreen MabSelect SuRe LX Columns, Cytiva
Chromatography Columns
This protein A column is prepacked with MabSelect SuRe LX affinity resin and is part of our process development platform. These packed columns are an excellent choice for method optimization and parameter screening for capture of monoclonal antibodies when a high dynamic binding capacity is needed.
  • The alkali-tolerant recombinant protein A ligand of MabSelect Sure LX allows clean-in-place (CIP) and the resin gives excellent dynamic binding capacity
  • Prepacked, ready-to-use columns for convenient chromatography process development
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening.
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography resin using the same linear fluid velocity
The small column volume of 4.7 ml and bed height of 10 cm make HiScreen columns excellent tools for method optimization, parameter screening, robustness testing, and convenient scale-up of the protein purification process. Process fluid velocities can be applied, since the 10 cm bed height gives enough residence time and the results can then serve as basis for linear process scale-up.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty